N-(3-Aminopropyl)cyclohexylamine

Supplier: SPECTRUM CHEMICAL MFG CORP

A1462-100ML
75871-706EA 212 USD
75871-706
N-(3-Aminopropyl)cyclohexylamine
N-(3-Aminopropyl)cyclohexylamine
Formula: C₉H₂₀N₂
Storage Temperature: Ambient
MDL Number: MFCD00003838
CAS Number: 3312-60-5
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR