Dipentyl ether

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: n-Pentyl etherDi-n-amyl etherAmyl ether

A1424-25ML A1424-100ML
75873-622EA 95 USD
75873-622 75871-678
Dipentyl ether
Dipentyl ether
Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009501
CAS Number: 693-65-2
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR