Pentylbenzene

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-PhenylpentaneAmylbenzene

A1422-100ML
75871-676EA 240.72 USD
75871-676
Pentylbenzene
Pentylbenzene
Formula: C₁₁H₁₆
MW: 148.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009502
CAS Number: 538-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR