Aminoacetonitrile sulfate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Cyanomethylamine sulphate
, Cyanomethylamine Sulfate
A1327-25GM
A1327-100GM
75871-702EA
85
USD
75871-702
75871-700
Aminoacetonitrile sulfate
Aminoacetonitrile sulfate
Formula:
C₂H₄N₂ MW: 210.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012851 CAS Number: 5466-22-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...