VWR® Critical Cover® SureGrip® Shoe Covers
Supplier: VWR International
VWR®
SH-12522-B SH-X1224-B SH-X1272-B SH-X1223-B SH-X1222-B SH-X1273-B SH-X1274-B SH-12523-B
10845-196CS 208.24 USD InStock
10845-196 47732-828 47732-830 47732-826 47732-824 47732-832 47732-836 10845-200
VWR® Critical Cover® SureGrip® Shoe Covers
Apparel Shoe and Boot Covers
Critical Cover® SureGrip® Shoe Covers have established themselves as the shoe cover of choice for a wide range of floor surfaces and in many different applications.
  • Combining durability
  • Anti-skid properties
  • SureGrip provides you with a blend of unmatched performance and overall value
Multiple style and packaging options add to the versatility of SureGrip, available in two styles, serged seam and conductive to meet any/all market applications.
Serged seams for added strength, traction and a snug fit for safety, unique conductive shoe cover for those environments where electro-static dissipation (ESD) protection is required. Non-skid, thread colors for convenient size identification, serged seams.
Gaylord packaging option (30 individual bags of 300 each per master carton) reduces waste, labor, and purchase orders while increasing your shelf-space and providing a reusable, pallet sized container for recyclables or other materials. Non-skid, thread colors for convenient size identification, serged seams.
Conductive for ESD concerns, anti-skid, serged seams.