LIT-927 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: 4-(4-Chlorophenyl)-6-(4-hydroxy-3-methoxyphenyl)pyrimidin-2(1H)-oneLIT927

AB287051-25MG AB287051-5MG
76325-500EA 972.93 USD
76325-500 76325-498
LIT-927 ≥98% (by HPLC)
LIT-927

LIT-927 is a locally and orally active CXCL12 neutraligand (Ki = 267 nM for CXCL12-TR binding). Displays anti-inflammatory effect in a murine model of allergic airway hypereosinophilia.


  • A potent and selective CXCL12 neutraligand

Formula: C₁₇H₁₃ClN₂O₃
MW: 328.75 g/mol
Storage Temperature: Freezer
CAS Number: 2172879-52-4
https://us.vwr.com/stibo/web/std.lang.all/87/90/27368790.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR