1,4-Dithioerythritol ≥99% (by titrimetric analysis)

Supplier: BioVision

Synonyms: Cleland's ReagentDL-Dithioerythritoltrans-1,4-Dimercaptobutane-2,3-diolDTEDithioerythritol(R*,S*)-1,4-Dimercaptobutane-2,3-diolErythro-1,4-Dimercaptobutane-2,3-diol

B2410-25G AB287037-1G AB287037-5G
76325-678EA 719.88 USD
76325-678 76325-680 76325-676
1,4-Dithioerythritol ≥99% (by titrimetric analysis)
1,4-Dithioerythritol

A useful reagent for maintaining sulfhydryl groups in the reduced state


  • A disulfide reducing agent

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063750
CAS Number: 92-68-8
https://us.vwr.com/stibo/web/std.lang.all/87/68/27368768.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR