Carbamazepine ≥98% (by HPLC)

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

B2401-250 B2401-50
76325-642EA 227.66 USD
76325-642 76325-640
Carbamazepine ≥98% (by HPLC)
Carbamazepine

Carbamazepine is an inhibitor of neuronal voltage-gated Na+ channels. It displays anticonvulsant activity and potentiates GABA-induced Cl- currents in HEK 293 cells expressing the GABAA receptor α1β2γ2 subtype combination. Can induce autophagy by inhibiting inositol synthesis. Also acts as an in vitro inducer of CYP3A4, the main human metabolizing enzyme present in the liver and intestine.


  • Inhibitor of neuronal NaV channels

Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005073
CAS Number: 298-46-4
https://us.vwr.com/stibo/web/std.lang.all/87/59/27368759.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR