Rotational Viscometer, ViscoQC 100
105020_US 105022_US 105021_US
76307-502EA 3897.81 USD
76307-502 76307-500 76307-498
Rotational Viscometer, ViscoQC 100
ViscoQC 100 is designed to bring simplicity to every operations for single-point viscosity determination.
  • Automatic spindle detection with magnetic coupling
  • Unique TruMode™ finds the right speed
  • Automatic spindle recognition for error-free measurements using Toolmaster™
  • Perfect traceability by automatic detection of attached L or R guard
  • Peltier temperature control with air counter-cooling for accurate sample temperature control (15 to 80 °C)
  • Prolonged spindle lifetime, high chemical resistance of stainless steel AISI 316
The unique air-counter-cooled Peltier temperature device for DIN spindles saves an enormous amount of space and time by providing highly accurate sample temperature control.
Monitor the sample temperature with a Pt100 sensor (–60 to +300 °C) and a flexible cup holder ensures the sample container is always centered.
Intuitive user interface on 3.5" color LCD display with predefined modes.Increase sample throughput using disposable aluminum cups (D18).
Certifications: Fulfills requirements of GMP, GAMP 5 (except 21 CFR Part 11).
Delivery information: A free installation and operational qualification document and Free V-Collect data collection software included; Extended three-year warranty program included; Relative spindles (L, RH) according to ISO 2555 included in standard delivery.