VWR® Horizontal MAXI E×W Gel Electrophoresis Systems
Supplier: VWR
VWR®
81-2325R
76314-862EA 599 USD
76314-862
VWR® Horizontal MAXI E×W Gel Electrophoresis Systems
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
VWR® MAXI E×WR model includes a buffer circulation system, powered by the electrodes themselves.
The hydrogen bubbles produced at the cathode pass up a diagonally positioned tube connected to the two sides of the tank producing a gentle circulation of the buffer. The result is a no pH and ionic gradient - without the need of mechanical pumps.
Delivery information: Includes integral buffer circulation, safety lid with retractable sheathed leads, UVT gel tray with gasketed end gates, four combs, two 25 well and two 50 well (1.5 mm thick).