MERC186
186-12
CA34172-040
MERC186
For the detection of amyloid deposits
  • For In-Vitro Diagnostic Use.
  • for Professional Use.
Used for cytoplasmic staining of paraffin sections fixed in Bouin’s fluid. Also a general tissue stain for use as a cytoplasmic counterstain.
Certifications: Certified by the Biological Stain Commission (BSC)