Wright's stain, BAKER ANALYZED®, Certified Stain for hematology, J.T.Baker®

Warning

Synonyms: Eosin Methylene blue solution according to WrightWright's eosin methylene blue

Management of Change (MOC) category = NI

BAKRX492-03 X492-03
JTX492-3EA 123.98 USD
JTX492-3 CAJTX492-3
Wright's stain, BAKER ANALYZED®, Certified Stain for hematology, J.T.Baker®
Wright's stain

Certified for use in blood staining.

Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
EINECS: 273-541-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Biological Test Passes Test
Certified by the Biological Stain Commission

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR