Trypsinizing Flasks with 3 Deep Side Baffles and 15-415 Sidearm, Bellco Glass
1989-00500 1989-00250 1989-00125
100483-156CS 365.19 USD
100483-156 100483-046 100483-154
Trypsinizing Flasks with 3 Deep Side Baffles and 15-415 Sidearm, Bellco Glass
Designed with deep side baffles for optimal agitation.
  • Type 1 Class A borosilicate glass
  • Promotes enzyme digestion of tissue fragments and homogenates into cell suspensions
Includes rubber lined, black phenolic screw caps. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense.