Toluidine blue O, HARLECO®, certified by the Biological Stain Commission stain for histology (CI 52040), Sigma-Aldrich®

Synonyms: TBO3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chlorideBlutene chlorideTolonium chlorideBasic Blue 17

364-12 364-12
34172-176EA 425.65 USD
CA34172-176 34172-176
Toluidine blue O, HARLECO®, certified by the Biological Stain Commission stain for histology (CI 52040), Sigma-Aldrich®
Toluidine blue O

For use in histological sections, animal tisuue and as a general nuclear stain.


Certified by the Biological Stain Commission (BSC)

Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011934
CAS Number: 92-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR