Methylene blue, Powder, HARLECO®, certified by the Biological Stain Commission stain for histology, stain for bacteriology, Sigma-Aldrich®
Synonyms:
Basic Blue 9
, Swiss Blue
, Methylthioninium chloride
, Methylene blue chloride
, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride
284-16
284-12
284-23
284-23
284-12
284-16
15204-114EA
953.32
USD
15204-114
CA15204-114
CA15204-112
34172-054
15204-112
CA34172-054
Methylene blue, Powder, HARLECO®, certified by the Biological Stain Commission stain for histology, stain for bacteriology, Sigma-Aldrich®
Methylene blue
Used for histological and bacteriological staining and as a constituent of Wright Stain.
Formula:
C₁₆H₁₈ClN₃S MW: 319.86 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...