Triethylammonium formate solution 1 M pH 6.0 volatile buffer, for HPLC, Fluka™

Warning

17901-500ML
BJ17901-500MLEA 489.1 USD
BJ17901-500ML
Triethylammonium formate solution 1 M pH 6.0 volatile buffer, for HPLC, Fluka™
Triethylammonium formate
Formula: C₇H₁₇NO₂
Density: 1.00 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134943
CAS Number: 585-29-5

Order Now

Specification Test Results

Density (D 20/4) 0.995 - 1.004
pH (25 °C) 5.9 - 6.1
Non-volatile matter Max. 0.05 %
Absorbance 280 nm Max. 0.05
Absorbance at 260 nm Max. 0.1
Filter test complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR