Sodium periodate ≥99.8% ACS, Sigma-Aldrich®
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Sodium metaperiodate
311448-5G
311448-100G
311448-500G
311448-5G
311448-500G
311448-2.5KG
311448-100G
BJ311448-5GEA
73.05
USD
BJ311448-5G
BJ311448-2.5KG
220054-854
220054-855
220054-856
BJ311448-100G
BJ311448-500G
Sodium periodate ≥99.8% ACS, Sigma-Aldrich®
Sodium periodate
Formula:
NaIO₄ MW: 213.89 g/mol Melting Pt: 270 °C Density: 3.86 g/cm3 Storage Temperature: Ambient |
MDL Number:
MFCD00003534 CAS Number: 7790-28-5 EINECS: 232-197-6 UN: 3085 ADR: 5.1,I |
Specification Test Results
Assay (excluding dried substances) | 99.8 - 100.3 % |
Manganese (Mn) | max. 3.0 ppm |
Other halogens (as Cl) | max. 0.02 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...