Sodium perchlorate ≥98.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

410241-100G 410241-2.5KG 410241-500G
BJ410241-500GEA 289.01 USD
BJ410241-500G BJ410241-2.5KG BJ410241-100G
Sodium perchlorate ≥98.0% ACS, Fluka™
Sodium perchlorate
Formula: ClNaO₄
MW: 122.44 g/mol
Melting Pt: 469 °C (dec.)
Density: 2.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003480
CAS Number: 7601-89-0
UN: 1502
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
pH (5 %, 25 °C) 6.0 - 8.0
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 0.02 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.05 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.003 %
Sulfate (SO₄) Max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR