Sodium hydrogen sulfate, anhydrous ∼95,0% (by titrimetric analysis), Purum, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Sodium bisulfate
, Sodium bisulphate
71656-250G
71656-1KG
71656-5KG
BJ71656-5KGEA
233.42
USD
BJ71656-5KG
BJ71656-1KG
BJ71656-250G
Sodium hydrogen sulfate, anhydrous ∼95,0% (by titrimetric analysis), Purum, Fluka™
Sodium hydrogen sulphate
Formula:
HNaO₄S MW: 120.06 g/mol Melting Pt: 315 °C Density: 2.12 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003531 CAS Number: 7681-38-1 EINECS: 231-665-7 |
Specification Test Results
Assay | 93.0 - 102.0 % |
Assay of Na₂SO₄ | Max. 6 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...