Sodium formate ≥99.0% (by non aqueous titration) for HPLC, Fluka™

Synonyms: Formic acid sodium salt

17841-250G 17841-50G
BJ17841-50GEA 97.57 USD
BJ17841-50G BJ17841-250G
Sodium formate ≥99.0% (by non aqueous titration) for HPLC, Fluka™
Sodium formate
Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 255 °C
Density: 1.92 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
pH (10 %, 20°C) 7.0 - 8.5
Colour of the solution complying
Turbidity of solution complying
Absorbance 500 nm Max. 0.02
Absorbance at 330 nm Max. 0.02
Absorbance at 300 nm Max. 0.03
Absorbance at 270 nm Max. 0.04
Absorbance at 260 nm Max. 0.05
Suitability test (HPLC) complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR