Potassium hexacyanoferrate(II) trihydrate ≥99%, puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Synonyms: Tetrapotassium hexacyanoferrate trihydratePotassiumhexacyanoferrate(II) trihydratePotassium ferrocyanide trihydrate

31254-6X500G 31254-1KG
BJ31254-500GCS 526.27 USD
BJ31254-500G BJ31254-1KG
Potassium hexacyanoferrate(II) trihydrate ≥99%, puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Potassium hexacyanoferrate (II) trihydrate
Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 70 °C
Density: 1.85 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2

Order Now

Specification Test Results

Assay Min. 99 %
Water insoluble matter Max. 0.005 %
Free alkali (as Carbonate) Max. 0.0015 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.002 %
Sodium (Na) Max. 0.01 %
Lead (Pb) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR