n-Dodecane ≥99%, Reagent Grade
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Dodecane
D221104-500ML
D221104-2.5L
D221104-100ML
BJD221104100MLEA
122.48
USD
BJD221104100ML
BJD221104500ML
BJD221104-2.5L
n-Dodecane ≥99%, Reagent Grade
n-Dodecane
Formula:
H₃C(CH₂)₁₀CH₃ MW: 170.34 g/mol Boiling Pt: 215-217 °C Melting Pt: -9.6 °C Density: 0.75 g/cm³ Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008969 CAS Number: 112-40-3 EINECS: 203-967-9 |
Specification Test Results
Vapor Pressure | 1 mmHg ( 47.8 °C) |
IR spectrum | complying |
Assay (GC) | Min. 99 % |
Refractive index (n 20/D) | 1.420 - 1.424 |
APHA | Max. 20 |
Vapor Density | 5.96 (vs air) |
Autoignition Temperature | 401 °F |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...