Lithium sulfate monohydrate, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

62609-500G 62609-100G
BJ62609-500GEA 194.69 USD
BJ62609-500G BJ62609-100G
Lithium sulfate monohydrate, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™
Lithium sulphate monohydrate
Formula: Li₂SO₄·H₂O
MW: 127.96 g/mol
Density: 2.060 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7

Order Now

Specification Test Results

Assay (excluding dried substance) 99.0 - 101.0 %
Water insoluble matter Max. 0.01 %
Loss on drying (150°C) 13.0 - 15.0 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 20 ppm
Nitrate (NO₃) Max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR