Lanthanum(III) nitrate hexahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

61520-100G 61520-500G
BJ61520-100GEA 231.45 USD
BJ61520-100G BJ61520-500G
Lanthanum(III) nitrate hexahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™
Lanthanum (III) nitrate hexahydrate
Formula: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Boiling Pt: 126 °C (Decomposition)
Melting Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 10 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 10 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 10 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR