Lanthanum(III) chloride heptahydrate ≥99%, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

31820-6X100G
BJ31820-100GCS 923.61 USD
BJ31820-100G
Lanthanum(III) chloride heptahydrate ≥99%, Puriss. p.a., Fluka™
Lanthanum (III) chloride heptahydrate
Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay Min. 99 %
Calcium (Ca) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.001 %
Magnesium (Mg) Max. 0.0005 %
Sodium (Na) Max. 0.001 %
Heavy metals (as Pb) Max. 0.0005 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR