Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Synonyms:
Glysanin
, Glycerin
, Glyceritol
, Propane-1,2,3-triol
, Osmoglyn
, Propanetriol
, Glycerol
, Glyrol
, Glycyl alcohol
, 1,2,3-Propanetriol
, Trihydroxypropane
49770-2.5L
49770-1L
49770-250ML
BJ49770-250MLEA
80.54
USD
BJ49770-250ML
BJ49770-1L
BJ49770-2.5L
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Glycerine
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Boiling Pt: 290 °C (1.013 hPa) Melting Pt: 18 °C Density: 1.26 g/cm³ (20 °C) Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 |
Specification Test Results
Assay (GC) | Min. 99.5 % |
Density (D 20/4) | 1.261 - 1.263 |
Refractive index (n 20/D) | 1.473 - 1.475 |
Water (Karl Fischer) | Max. 0.1 % |
Residue on ignition | Max. 0.005 % |
Indifference | complying |
Calcium (Ca) | Max. 0.0005 % |
Cadmium (Cd) | Max. 0.0001 % |
Cobalt (Co) | Max. 0.0001 % |
Chromium (Cr) | Max. 0.0001 % |
Copper (Cu) | Max. 0.0001 % |
Iron (Fe) | Max. 0.0001 % |
Potassium (K) | Max. 0.002 % |
Magnesium (Mg) | Max. 0.0001 % |
Manganese (Mn) | Max. 0.0001 % |
Sodium (Na) | Max. 0.002 % |
Nickel (Ni) | Max. 0.0001 % |
Lead (Pb) | Max. 0.0001 % |
Zinc (Zn) | Max. 0.0001 % |
Heavy metals (as Pb) | Max. 0.0002 % |
Halogene compounds (as Cl) | Max. 0.003 % |
Ammonium (NH₄) | Max. 0.0005 % |
Chloride (Cl) | Max. 0.0001 % |
Sulfate (SO₄) | Max. 0.001 % |
Reaction against H₂SO₄ | complying |
Acrolein, Glucose + NH4-compounds | complying |
Fatty acids and ester (as butyric acid) | Max. 0.05 % |
APHA | Max. 10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...