Eriochrome cyanine R S. No. 840, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Chromoxane Cyanine R

32752-25G
BJ32752-25GEA 72.57 USD
BJ32752-25G
Eriochrome cyanine R S. No. 840, Fluka™
Eriochrome cyanine R
Formula: C₂₃H₁₅Na₃O₉S
Storage Temperature: Ambient
MDL Number: MFCD00012411
CAS Number: 3564-18-9
EINECS: 222-641-7

Order Now

Specification Test Results

Identity (IR) complying
Suitability complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR