EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Ethylendiamintetraacetic acid magnesium dipotassium salt
33350-6X100G
33350-50KG
BJ33350-50KGEA
13067.47
USD
BJ33350-50KG
BJ33350-100G
EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™
EDTA magnesium dipotassium salt
Formula:
C₁₀H₁₂K₂MgN₂O₈ Storage Temperature: Ambient |
MDL Number:
MFCD00054450 CAS Number: 15708-48-2 EINECS: 239-803-8 |
Specification Test Results
Assay | Min. 98 % |
pH (0.1 M, 20°C) | 8.5 - 9.5 |
Calcium (Ca) | Max. 0.05 % |
Iron (Fe) | Max. 0.001 % |
Heavy metals (as Pb) | Max. 0.001 % |
Chloride (Cl) | Max. 0.005 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...