Cresol red ≥83% indicator, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
, o-Cresolsulfonphthalein
, o-Cresolsulphonphthalein
32653-25G
32653-10G
BJ32653-25GEA
185.61
USD
BJ32653-25G
BJ32653-10G
Cresol red ≥83% indicator, Fluka™
Cresol red
Formula:
C₂₁H₁₈O₅S MW: 382.43 g/mol Density: 0.998 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00005878 CAS Number: 1733-12-6 EINECS: 217-064-2 |
Specification Test Results
Identity (IR) | complying |
Assay (HPLC) | Min. 83 % |
Loss on drying | Max. 5 % |
Change range (pH 0.5-2.5) | complying |
Change range (pH 6.5-8.5) | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...