Calcein disodium salt for complexometry, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Calcein sodium salt

21030-5G 21030-100G
BJ21030-100GEA 969.61 USD
BJ21030-100G BJ21030-5G
Calcein disodium salt for complexometry, Fluka™
Calcein disodium salt
Formula: C₃₀H₂₄N₂Na₂O₁₃
Storage Temperature: Ambient
MDL Number: MFCD00082336
CAS Number: 108750-13-6

Order Now

Specification Test Results

Appearance of the substance complying
Identity (IR) complying
Suitability as indicator complying
Absorption maximum 494 - 504 nm
Absorbtion (1%/1cm) Min. 600
Absorbtion (1%/1cm) min. 600

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR