Cadmium(II) chloride, anhydrous ≥99.0%, puriss. p.a.

Danger

Synonyms: Cadmium chloride

20899-25G
BJ20899-25GEA 62.64 USD
BJ20899-25G
Cadmium(II) chloride, anhydrous ≥99.0%, puriss. p.a.
Cadmium (II) chloride
Formula: CdCl₂
MW: 183.32 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 356 - 409 °C
Density: 3.91 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 100 ppm
Copper (Cu) Max. 10 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 200 ppm
Sodium (Na) Max. 500 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 500 ppm
Total nitrogen (as NO₃) Max. 0.003 %
Ammonium (NH₄) Max. 100 ppm
Sulfate (SO₄) Max. 100 ppm
Vapor Pressure 10 mmHg ( 656 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR