Barium carbonate ≥99%, Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

11729-1KG
BJ11729-1KGEA 219.01 USD
BJ11729-1KG
Barium carbonate ≥99%, Puriss. p.a. ACS, Fluka™
Barium carbonate
Formula: BaCO₃
MW: 197.34 g/mol
Melting Pt: 1.400 °C
Density: 4.4 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Water-soluble base Max. 0.002 meq/g
Insoluble in HCl Max. 0.015 %
Calcium (Ca) Max. 500 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 7000 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 20 ppm
Sulfide (S) Max. 10 ppm
Oxidizing substances (as NO₃) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR