Ammonium cerium(IV) nitrate ≥98.5% (RT), puriss. p.a. ACS, Fluka™

Danger

Synonyms: Ammonium hexanitratocerate (IV)Ammonium ceric nitrateCerium (IV) ammonium nitrateCeric Ammonium NitrateDiammonium hexanitratocerate

22249-500G 22249-100G
BJ22249-500GEA 348.26 USD
BJ22249-500G BJ22249-100G
Ammonium cerium(IV) nitrate ≥98.5% (RT), puriss. p.a. ACS, Fluka™
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 214 °C
Density: 2,49 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 3085
ADR: 5.1 (8),II

Order Now

Specification Test Results

Assay 98.5 - 101.0 %
Insoluble in acid Max. 0.05 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 10 ppm
Magnesium (Mg) Max. 20 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 10 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR