Ammonium bromide ≥99.0% ACS, Fluka™

Warning

213349-500G 213349-2.5KG
BJ213349-500GEA 112.91 USD
BJ213349-500G BJ213349-2.5KG
Ammonium bromide ≥99.0% ACS, Fluka™
Ammonium bromide
Formula: H₄BrN
MW: 97.94 g/mol
Melting Pt: 452 °C
Density: 2.43 g/cm3 (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8

Order Now

Specification Test Results

Vapor Pressure 1 mmHg ( 198.3 °C)
Assay 99.0 - 101.0 %
pH (5 %, 25 °C) 4.8 - 6.0
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.01 %
Barium (Ba) Max. 0.002 %
Iron (Fe) Max. 5 ppm
Heavy metals (ICP) ACS Max. 5 ppm
Bromate (BrO₃) Max. 0.002 %
Chloride (Cl) Max. 0.2 %
Iodide (I) test complying
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR