Aluminium potassium sulfate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Aluminium Potassium sulphate, 12-Hydrate
, Aluminum potassium sulphate dodecahydrate
, Alum potassium
, Aluminium Potassium sulfate, 12-Hydrate
, Potassium alum
, Aluminium potassium bis(sulfate) dodecahydrate
, Aluminum potassium sulfate dodecahydrate
, Aluminium potassium bis(sulphate) dodecahydrate
12623-6X1KG
BJ12623-1KGCS
1220.14
USD
BJ12623-1KG
Aluminium potassium sulfate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Aluminium potassium sulphate dodecahydrate
Formula:
AlK(SO₄)₂·12H₂O MW: 474.39 g/mol Melting Pt: 92.5 °C Density: 1.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149143 CAS Number: 7784-24-9 |
Specification Test Results
Assay (calculated to the dried substance) | 99.0 - 100.5 % |
Loss on drying | 43.0 - 46.0 % |
Ammonium (NH₄) | Max. 0.2 % |
Iron (Fe) | complying |
Arsenic (As) | Max. 0.0002 % |
Heavy metals (as Pb) | Max. 0.001 % |
Chloride (Cl) | Max. 0.005 % |
Appearance of the solution | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...