Aluminium chloride hexahydrate ≥99.0% (by argentometric titration), crystallised, purum p.a.

Warning

Synonyms: Aluminum chloride hexahydrate

06232-1KG 06232-250G
BJ06232-1KGEA 120.91 USD
BJ06232-1KG BJ06232-250G
Aluminium chloride hexahydrate ≥99.0% (by argentometric titration), crystallised, purum p.a.
Aluminium chloride hexahydrate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: AlCl₃·6H₂O
MW: 241.43 g/mol
Density: 2,4 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
pH (5 %, 25 °C) 2.5 - 3.5
Water (Karl Fischer) 42 - 48 %
Calcium (Ca) Max. 100 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 100 ppm
Sodium (Na) Max. 500 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Sulfur (S) Max. 15 ppm
Total S (as SO₄) Max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR