2,6-Dichlorophenolindophenol sodium salt hydrate ≥90% (dry basis) redox indicator, Fluka™

Synonyms: Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide hydrateDPIP2,6-Dichlorophenolindophenol sodium salt xhydrateDCIPSodium 2,6-dichloroindophenolate hydrate2,6-Dichloroindophenol, sodium salt hydrate2,6-Dichloroindophenol sodium hydrate2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt hydrateTillmans' reagent hydrate

33125-25G 33125-5G
BJ33125-25GEA 305.09 USD
BJ33125-25G BJ33125-5G
2,6-Dichlorophenolindophenol sodium salt hydrate ≥90% (dry basis) redox indicator, Fluka™
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula: C₁₂H₆Cl₂NNaO₂·xH₂O
MW: 290.08 g/mol (anhy)
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150014
CAS Number: 620-45-1
EINECS: 210-640-4

Order Now

Specification Test Results

Assay (calculated to the dried substance) Min. 90 %
Loss on drying (120°C) Max. 14 %
Foreign dye stuffs complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR