1-Octanol ≥99% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
n-Octanol
472328-4L
472328-100ML
472328-1L
472328-2.5L
BJ472328-2.5LEA
229.73
USD
BJ472328-2.5L
BJ472328-4L
BJ472328-1L
BJ472328-100ML
1-Octanol ≥99% ACS, Riedel-de Haën™
1-Octanol
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 194 °C (1.013 hPa) Melting Pt: -15 °C Density: 0.827 g/cm³ (25 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 |
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 99.0 % |
Non-volatile matter | Max. 0.004 % |
Titrable Acid | Max. 0.0002 meq/g |
APHA | Max. 10 |
Refractive Index | n20/D 1.429 (lit.) |
Vapor Pressure | 0.14 mmHg ( 25 °C) |
Autoignition Temperature | 523 °F |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...