Curly Haired Tarantula
Supplier: VWR
Ward's®
WARD470312-394
470312-394EA 49.95 USD
470312-394
Curly Haired Tarantula
Specimens Live Specimens Animals - Living Material
Your tarantula is shipped in a plastic container with sphagnum moss as cushioning. It should be placed in a prepared habitat upon arrival.
  • Brachypelma albopilosa
Ordering information: These products contain living or perishable materials. Visit https://wardsci.com/cms/teacher_live_materials for care instructions.
Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.