Chlortetracycline hydrochloride, crystals

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 7-Chlorotetracycline hydrochlorideAureomycin

C1601-5GM C1601-100GM
75872-104EA 333.05 USD
75872-104 75872-108
Chlortetracycline hydrochloride, crystals
Chlortetracycline hydrochloride

Chlortetracycline Hydrochloride, Crystal | Non-graded Chlortetracycline Hydrochloride, Crystal supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₂H₂₃ClN₂O₈·HCl
MW: 515.34 g/mol
Melting Pt: 220°C/428°F
Storage Temperature: Freezer
MDL Number: MFCD00082440
CAS Number: 64-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR