SatPax® MicroPolx® Sterile Nonwoven 100% Polyester Microfiber Wet-Laid Cleanroom Wipes, Saturated, Berkshire
Supplier: Berkshire
SatPax®
SSPMPNW500124
76318-648CS 603.63 USD
76318-648
SatPax® MicroPolx® Sterile Nonwoven 100% Polyester Microfiber Wet-Laid Cleanroom Wipes, Saturated, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
MicroPolx® nonwoven cleanroom validated sterile wipes, a cutting edge innovation highlighting an inventive assembling process of 100% polyester microfiber wet-laid nonwoven material.
  • Extremely soft hand
  • Mechanical strength
  • Anti-scratch
  • Multi–directional layering
  • Gamma irradiated
  • Lot to lot traceability
Wipes clean sensitive surfaces without damage while maximizing particle pickup and retention. Superior microfiber contamination removal performance.
Certifications: Designed for use in ISO class 5 and higher cleanroom environments.