Sudan III in isopropanol (IPA) fat and lipid stain
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Sudan Red III
, D and C Red No. 17
, Sudan Red BK
, 1-(4-(Phenylazo)phenylazo)-2-naphthol
, Sudan® III (Red)
, Sudan G
, Cerasin Red
, Tony Red
, Oil scarlet
, Scarlet B
, Organol® red BS
, Fat Ponceau G
, Oil red
, Solvent Red 23
, Sudan Red B
, 1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-ol
, Sudan V
26503-03
100504-656EA
40.14
USD
100504-656
Sudan III in isopropanol (IPA) fat and lipid stain
Sudan III
For fat and lipid stain.
Stain results for Fat: Deep orange-red with Oil Red O, lighter with Sudan IV, Orange-yellow with Sudan II
Formula:
C₂₂H₁₆N₄O MW: 352.4 g/mol Flash Pt: 13 °C (55.4 °F) |
MDL Number:
MFCD00003905 CAS Number: 85-86-9 UN: 1219 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...