Gallego's differentiating solution for bacteriology (Brown-Hopps method)
Supplier: Electron Microscopy Sciences
Danger
26106-04
100504-502EA 30.2 USD
100504-502
Gallego's differentiating solution for bacteriology (Brown-Hopps method)
Gallego's differentiating solution
Brown-Hopps Method reagent for Gram Positive and Gram Negative Bacteria.
Stain results:
Gram Negative Bacteria: Red
Gram Positive Bacteria: Blue
Background: Yellow