6-Carboxy-X-rhodamine triethylammonium salt single isomer ≥95% (by TLC) amine-reactive fluorescent dye
Supplier: Biotium
Synonyms: 6-ROXCarboxy-X-rhodamine triethylammonium salt, 6-isomer
90007
89139-528EA 168.56 USD
89139-528
6-Carboxy-X-rhodamine triethylammonium salt single isomer ≥95% (by TLC) amine-reactive fluorescent dye
6-Carboxy-X-rhodamine triethylammonium salt
6-ROX (6-Carboxy-X-rhodamine) is a single isomer, long wavelength rhodamine dye.
  • λEx/λ Em(MeOH) = 568/595 nm
  • Dark red solid soluble in DMSO, DMF or MeOH
  • Store at 4°C and protect from light