Guanidinium hydrochloride, Biotech Reagent, J.T.Baker®

Warning

Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideGuanadine hydrochloride

Management of Change (MOC) category = R

BAKR0510-06 BAKR0510-F9 BAKR0510-F9
JT0510-F9CS 36668.87 USD
CAJT0510-F9 CAJT0510-06 JT0510-F9
Guanidinium hydrochloride, Biotech Reagent, J.T.Baker®
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://us.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(:NH)NH₂·HCl) 99.5 - 101.0 %
Acidity (as HCl) ≤0.01 %
Loss on Drying at 105°C ≤0.5 %
Residue after Ignition ≤0.050 %
Trace Impurities - Copper (Cu) ≤0.200 ppm
Heavy Metals (as Pb) ≤0.5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Enzyme Activity - DNase Activity Passes Test
Enzyme Activity - RNase Activity Passes Test
Enzyme Activity - Protease Activity Passes Test
Absorbance of a 6M Aqueous Solution - 230 nm ≤0.200
Absorbance of a 6M Aqueous Solution - 260 nm ≤0.030
Absorbance of a 6M Aqueous Solution - 275 nm ≤0.030
Arsenic and Antimony (as As) ≤0.500 ppm
Nitrate (NO₃) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Cyanide (CN) ≤0.1 ppm
Particulate Matter Passes Test
Water-Insoluble Matter ≤0.3 %
Melamine and Related Compounds Passes Test
Endotoxin, EU/mL, For Information Only
Total Aerobic Plate Count, For Information Only
Total Yeast and Mold, For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Metallic Impurities - Potassium (K), For Information Only
Metallic Impurities - Sodium (Na), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Lithium (Li), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Non-Metallic Impurities - Arsenic (As), For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR