VWR® Hazardous Series Refrigerator and Freezers
Supplier: VWR, part of Avantor
50FREEVWA
76314-286EA 10734.46 USD
76314-286
VWR® Hazardous Series Refrigerator and Freezers
VWR® Hazardous series flammable laboratory refrigerator, designed to be used for the storage of volatiles, compounds, and solvents. Unit has manual defrost.
  • Spark free interior to reduce the risk of internal explosion
  • Designed to meet NFPA 45 and 99 along with OSHA article 1910.307
CFC- and HCFC-free refrigeration system and insulation with manual defrost.
Certifications: UL listed.