Masterflex® L/S® Precision BioPharm Plus Silicone (Platinum-Cured) Pump Tubing, Cole Parmer
Supplier: Cole Parmer
96440-25 96440-16
MSPP-96440-16EA 191 USD
MSPP-96440-16 MSPP-EW9644025
Masterflex® L/S® Precision BioPharm Plus Silicone (Platinum-Cured) Pump Tubing, Cole Parmer
Tubing Transfer Tubing
BioPharm Plus Silicone (Platinum-Cured) tubing for purity, flexibility, and longer tubing life.
  • Provides longer life than other platinum and peroxide silicone tubing formulations
  • Manufactured according to Good Manufacturing Practices (GMP)
  • Excellent heat resistance, low compression set, odorless, nontoxic and translucent
  • Ideal for pharmaceutical, biotechnology, peristaltic pumping, and food applications
  • Longer tubing life means less tubing changes and maintenance requirements
Ideal for pharmaceutical and biotechnological applications. Based on your application needs choose precision tubing or high-performance precision tubing. High-Performance precision tubing has a thicker wall that improves pressure generation suction lift, and the ability to pump viscous fluids. BioPharm Plus Silicone (platinum-cured) tubing has a working temperature range of –75 to 450 °F (–60 to 232 °C). Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide.
Certifications: This tubing exceeds USP class VI, 3A sanitary standards and is FDA compliant.