Nitric acid fuming ≥98%

Supplier: Puritan Products, Inc.
Danger

2712964
76266-904EA 1354.23 USD
76266-904
Nitric acid fuming ≥98%
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2032
ADR: 8,5.1,I

Order Now

Specification Test Results

Decapsulation Grade
Assay Max. 98%
Oxides of Nitrogen Max. 2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR