Calcium citrate tetrahydrate

Supplier: Pfaltz & Bauer
Warning

Synonyms: Calcium citrate tribasic tetrahydratetri-Calcium dicitrate tetrahydrate

C01125-500G
101185-078EA 230.15 USD
101185-078
Calcium citrate tetrahydrate
Calcium citrate tetrahydrate
Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
MW: 570.5 g/mol
MDL Number: MFCD00150786
CAS Number: 5785-44-4
EINECS: 212-391-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR