C-Flex® Standard Tubing, Cole Palmer
Supplier: Cole Parmer
06424-74
76300-154PK 174.02 USD
76300-154
C-Flex® Standard Tubing, Cole Palmer
Tubing Transfer Tubing
This tubing is mainly used for the general-purpose laboratory uses requiring high performance and long life.
  • Temperature range: –50 to +275 °F (–45 to +135 °C)
  • Sterilize with autoclave, ethylene oxide, or gamma irradiation
Features with characteristics such as exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability and a smooth surface.
Certifications: Resin meets USP Class VI requirements and FDA, European Pharmacopoeia, Non-DEHP, NSF-61, ADCF, 3A, Non-Phthalate, NSF-51 certified. Tubing is manufactured according to GMP.